Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2-formylpyridine
4-Bromo-2-formylpyridine 97%
4-Bromo-2-formylpyridine 97%
4-Bromo-2-formylpyridine 97%
APOSOR302555-1G
:
Apollo Scientific
:
4-Bromo-2-formylpyridine 97%
APOSOR302555-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
186.01
:
GHS07
:
Ambient
:
131747-63-2
:
C₆H₄BrNO
:
MFCD08690697