6-Bromo-1,2,3,4-tetrahydroquinoline
Catalog # APOSOR301104-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.0864 g/mol
Storage Temperature:Ambient
CAS Number:22190-35-8
Formula:C₉H₁₀BrN
MDL Number:MFCD04966989