Order Entry
Northern Ireland
ContactUsLinkComponent
Ethyl-3-cyclopropyl-3-oxopropanoate 95%
Irritant
Ethyl-3-cyclopropyl-3-oxopropanoate 95%
Catalog # APOSOR301083-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-3-cyclopropyl-3-oxopropanoate 95%
Catalog # APOSOR301083-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :156.18 g/mol
Boiling Pt:80…82 °C/6mmHg
Density:1,058 g/cm³ (25 °C)
Flash Pt:>96 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:24922-02-9
Formula:C₈H₁₂O₃
MDL Number:MFCD00184570