
1H-Pyrazol-5-ol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:60456-93-1
:137-45-1
Formula:C₃H₄N₂O
MDL Number:MFCD11227225
:84.08 g/mol
Melting Pt:169…179 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together