
3,5-Dinitrosalicylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:609-99-4
:
Formula:C₇H₄N₂O₇
MDL Number:MFCD00007104
:228.12 g/mol
Melting Pt:169…173 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together