Order Entry
Northern Ireland
ContactUsLinkComponent
S,S'-Dimethyl-N-cyanodithioiminocarbonate 97%
Corrosive
Irritant
S,S'-Dimethyl-N-cyanodithioiminocarbonate 97%
Catalog # APOSOR28666-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
S,S'-Dimethyl-N-cyanodithioiminocarbonate 97%
Catalog # APOSOR28666-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :146.23 g/mol
Melting Pt:45…50 °C
Flash Pt:>113 °C
Storage Temperature:Refrigerator
EINECS:233-467-6
UN:1759
ADR:8,II
 :GHS05
 :GHS07
CAS Number:10191-60-3
Formula:C₄H₆N₂S₂
MDL Number:MFCD00009825