
Coniferyl alcohol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:458-35-5
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00002922
:180.20 g/mol
Boiling Pt:163…165 °C/3mmHg
Melting Pt:75…80 °C
Storage Temperature:Freezer
EINECS:207-277-9
:GHS07
Frequently Bought Together