ContactUsLinkComponent
ω-Bromo-4-nitroacetophenone 98%
ω-Bromo-4-nitroacetophenone 98%
  APOSOR24069-25G
 :  
ω-Bromo-4-nitroacetophenone 98%
  APOSOR24069-25G
 :  
  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:
 : α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone
 :
 :244.04 g/mol
 :202-789-9
 :3261
 :GHS05
 :Ambient
 :99-81-0
 :C₈H₆BrNO₃
 :MFCD00007356
 :94…101 °C
 :8 II