
2-Iodobenzyl alcohol
Catalog # APOSOR2334-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:234.04 g/mol
Melting Pt:89…92 °C
EINECS:225-933-2
:GHS07
Storage Temperature:Ambient
CAS Number:5159-41-1
Formula:C₇H₇IO
MDL Number:MFCD00004610