
4-Bromo-o-phenylenediamine 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1575-37-7
:
Formula:C₆H₇BrN₂
MDL Number:MFCD02660622
Melting Pt:65…69 °C
UN:2811
ADR:6.1,III
:187.04 g/mol
:GHS06
Storage Temperature:Ambient
Frequently Bought Together