Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
(R)-(-)-1,3-Butanediol

(R)-(-)-1,3-Butanediol

Bottle, 1 g
Catalog # APOSOR2223-1G
Irritant
Supplier:   Apollo Scientific
(R)-(-)-1,3-Butanediol
Catalog # APOSOR2223-1G
Supplier:  Apollo Scientific
CAS Number:  6290-03-5
(R)-(-)-1,3-Butanediol

Bottle, 1 g

CAS Number:  6290-03-5
Price
£19.70
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (R)-(-)-1,3-Dihydroxy butane, (R)-(-)-1,3-Butylene glykol, (R)-Butane-1,3-diol
  • CAS Number:6290-03-5
  • Formula:C₄H₁₀O₂
  • MDL Number:MFCD00064277
  • Molecular Weight:90.12 g/mol
  • Boiling Pt:203…204 °C
  • Density:1,005 g/cm³ (25 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.