Order Entry
Northern Ireland
ContactUsLinkComponent
3-(Chlorosulphonyl)benzoic acid
3-(Chlorosulphonyl)benzoic acid
Catalog # APOSOR21680-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(Chlorosulphonyl)benzoic acid
Catalog # APOSOR21680-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :220.63 g/mol
Melting Pt:128…133 °C
Storage Temperature:Ambient
EINECS:223-692-8
UN:3261
ADR:8,II
 :GHS05
CAS Number:4025-64-3
Formula:C₇H₅ClO₄S
MDL Number:MFCD00024877