Order Entry
Northern Ireland
ContactUsLinkComponent
6-Methoxy-o-cresol 98%
Irritant
6-Methoxy-o-cresol 98%
Catalog # APOSOR21227-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Methoxy-o-cresol 98%
Catalog # APOSOR21227-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.17 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2896-67-5
Formula:C₈H₁₀O₂
MDL Number:MFCD00043771