(3-(Methylamino)propyl)trimethoxysilane
Catalog # APOSOR18841-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:193.32 g/mol
Boiling Pt:106 °C
Density:0,975 g/cm³ (25 °C)
Flash Pt:82 °C
EINECS:221-334-5
:GHS07
Storage Temperature:Ambient
CAS Number:3069-25-8
Formula:H₃CNHCH₂CH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00048021