
4-Chloro-3,5-dinitrobenzoic acid
Catalog # APOSOR18359-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:246.56 g/mol
Boiling Pt:315 °C
Melting Pt:159…162 °C
Flash Pt:186 °C
Storage Temperature:Ambient
EINECS:204-209-1
:GHS07
CAS Number:118-97-8
Formula:C₇H₃ClN₂O₆
MDL Number:MFCD00007080