
2-Bromonaphthalene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:580-13-2
:
Formula:C₁₀H₇Br
MDL Number:MFCD00004051
Boiling Pt:281…282 °C
Melting Pt:52…55 °C
:207.07 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together