Order Entry
Northern Ireland
ContactUsLinkComponent
4-Boronobenzenesulphonamide pinacol ester 98%
Irritant
4-Boronobenzenesulphonamide pinacol ester 98%
Catalog # APOSOR16633-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Boronobenzenesulphonamide pinacol ester 98%
Catalog # APOSOR16633-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :283.1516 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:214360-51-7
Formula:C₁₂H₁₈BNO₄S
MDL Number:MFCD06657806