
3-(2-Chlorophenyl)-1H-pyrazole
Catalog # APOSOR16-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.62 g/mol
Melting Pt:76…77 °C
:GHS07
Storage Temperature:Ambient
CAS Number:59843-55-9
Formula:C₉H₇ClN₂
MDL Number:MFCD00111882