Order Entry
Northern Ireland
ContactUsLinkComponent
5-Bromo-2-hydrazino-1H-1,3-benzimidazole 95%
Irritant
5-Bromo-2-hydrazino-1H-1,3-benzimidazole 95%
Catalog # APOSOR15310-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2-hydrazino-1H-1,3-benzimidazole 95%
Catalog # APOSOR15310-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.07 g/mol
Melting Pt:96…98 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:54303-63-8
Formula:C₇H₇BrN₄
MDL Number:MFCD08276936