Order Entry
Northern Ireland
ContactUsLinkComponent
2,2':6',2''-Terpyridine 97%
2,2':6',2''-Terpyridine 97%
Catalog # APOSOR13730-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2':6',2''-Terpyridine 97%
Catalog # APOSOR13730-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.27 g/mol
Boiling Pt:370 °C
Melting Pt:89…92 °C
Storage Temperature:Ambient
EINECS:214-559-5
UN:2811
ADR:6.1,I
 :GHS06
CAS Number:1148-79-4
Formula:C₁₅H₁₁N₃
MDL Number:MFCD00006213