
Ethyl-4-pyrazolecarboxylate 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:37622-90-5
:
Formula:C₆H₈N₂O₂
MDL Number:MFCD00010844
:140.14 g/mol
Boiling Pt:138…140 °C/3mmHg
Melting Pt:78…80 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together