
tert-Butyl-3-phenylpiperazine-1-carboxylate
Catalog # APOSOR12887-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:262.35 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:502649-25-4
Formula:C₁₅H₂₂N₂O₂
MDL Number:MFCD03840082