N-Boc-1,3-propanediamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75178-96-0
:
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD00210021
:174.244 g/mol
Boiling Pt:203 °C
Melting Pt:22 °C
Density:0,998 g/cm³ (25 °C)
Flash Pt:109 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together