Order Entry
Northern Ireland
ContactUsLinkComponent
1-Acetyladamantane 98%
Irritant
1-Acetyladamantane 98%
Catalog # APOSOR1265-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Acetyladamantane 98%
Catalog # APOSOR1265-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :178.28 g/mol
Melting Pt:53…55 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1660-04-4
Formula:C₁₂H₁₈O
MDL Number:MFCD00074739