Order Entry
Northern Ireland
ContactUsLinkComponent
2',4'-Dichloroacetophenone
Corrosive
Irritant
2',4'-Dichloroacetophenone
Catalog # APOSOR11049-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2',4'-Dichloroacetophenone
Catalog # APOSOR11049-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :189.04 g/mol
Boiling Pt:245…247 °C
Melting Pt:31…34 °C
Density:1,32 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:218-780-8
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:2234-16-4
Formula:C₈H₆Cl₂O
MDL Number:MFCD00000581