


1,3-Dichloro-5-nitrobenzene
Catalog # APOSOR1083-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:192 g/mol
Melting Pt:62…65 °C
EINECS:210-557-3
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:618-62-2
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD00007211