Order Entry
Northern Ireland
ContactUsLinkComponent
4-(Methylsulphonylamino)benzeneboronic acid 95%
4-(Methylsulphonylamino)benzeneboronic acid 95%
Catalog # APOSOR10562-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Methylsulphonylamino)benzeneboronic acid 95%
Catalog # APOSOR10562-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.03 g/mol
Melting Pt:148…154 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:380430-57-9
Formula:C₇H₁₀BNO₄S
MDL Number:MFCD02179473