Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Acetylthiophene
2-Acetylthiophene 97%
2-Acetylthiophene 97%
APOSOR10140-100G
:
Apollo Scientific
:
2-Acetylthiophene 97%
APOSOR10140-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
126.18 g/mol
:
214 °C
:
44510 °C
:
1,168 g/cm³ (25 °C)
:
91 °C
:
Refrigerator
:
201-804-6
:
2810
:
6.1,II
:
GHS06
:
88-15-3
:
C₆H₆OS
:
MFCD00005442