Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-b]pyridazine
Imidazo[1,2-b]pyridazine 98%
Imidazo[1,2-b]pyridazine 98%
APOSOR01790-25G
:
Apollo Scientific
:
Imidazo[1,2-b]pyridazine 98%
APOSOR01790-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
119.12 g/mol
:
53…54 °C
:
GHS07
:
Ambient
:
766-55-2
:
C₆H₅N₃
:
MFCD07782103