(S)-(+)-Camptothecin
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7689-03-4
:
Formula:C₂₀H₁₆N₂O₄
MDL Number:MFCD00081076
:348.36 g/mol
Melting Pt:260 °C (dec.)
Storage Temperature:Refrigerator
UN:1544
ADR:6.1,III
:GHS06
Frequently Bought Together