Zirconium(IV) ethoxide
Catalog # APOSIN3916-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:271.47 g/mol
Boiling Pt:235 °C/5mmHg
Melting Pt:171…173 °C
Storage Temperature:Ambient
CAS Number:18267-08-8
Formula:C₈H₂₀O₄Zr
MDL Number:MFCD00015654