



Sodium periodate 99%
:7790-28-5
:
:NaIO₄
:MFCD00003534
:300 °C (dec.)
:4,174 g/cm³ (25 °C)
:Ambient
:232-197-6
:3085
:5.1+8,I
:213.89 g/mol
:GHS03
:GHS05
:GHS08
:GHS09
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Iodine ≥99.5%, crystalline
: Thermo Fisher Scientific
Sodium sulphate, anhydrous 99%
: Thermo Fisher Scientific
Potassium chloride ≥99%
: Thermo Fisher Scientific
Trimethyloxonium tetrafluoroborate 95%
: Apollo Scientific