Order Entry
Northern Ireland
ContactUsLinkComponent
Tin(IV) tert-butanolate 99.99%
Toxic
Tin(IV) tert-butanolate 99.99%
Catalog # APOSIN3278-10G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Tin(IV) tert-butanolate 99.99%
Catalog # APOSIN3278-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Melting Pt:45 °C
Density:1,06 g/cm³ (25 °C)
Storage Temperature:Ambient
ADR:6.1,III
 :411.165 g/mol
UN:3146
 :GHS06
CAS Number:36809-75-3
Formula:C₁₆H₃₆O₄Sn
MDL Number:MFCD00156551