
Praseodymium trichloride, anhydrous
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10361-79-2
:
Formula:Cl₃Pr
MDL Number:MFCD00011176
:247.27 g/mol
Boiling Pt:1700 °C
Melting Pt:786 °C
Density:4,02 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together