Iron(II) sulphate heptahydrate ACS
Catalog # APOSIN2086-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:278.02 g/mol
Melting Pt:64 °C
Density:1,898 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:7782-63-0
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719