Holmium trichloride hexahydrate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14914-84-2
:
Formula:Cl₃Ho·6H₂O
MDL Number:MFCD00149739
:379.38 g/mol
Melting Pt:165 °C (dec.)
Storage Temperature:Ambient
Frequently Bought Together
Copper(II) sulphate, anhydrous ≥99.0%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Zinc chloride 98.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Calcium chloride dihydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JPC, FCC, E511, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
DISHWASHER SALT 2KG BAG GRANULAR 1 * 2 kg
Supplier: LAB3 OWN PRODUCTION
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Copper(II) sulphate pentahydrate 99.0-101.0%, crystallised Ph. Eur.
Supplier: VWR Chemicals
Potassium hydrogen phthalate 99.95-100.05% ACS primary standard
Supplier: Thermo Fisher Scientific