
Gadolinium trichloride, anhydrous 99,9%
:10138-52-0
:
:Cl₃Gd
:MFCD00011024
:263.61 g/mol
:609 °C
:2,424 g/cm³ (25 °C)
:233-386-6
:GHS07
:Ambient
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium dihydrogen phosphate ≥98%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Sodium bisulphite 50% (w/v) in aqueous solution, TECHNICAL
: VWR Chemicals
Citric acid, anhydrous ≥99.5%, powder, GPR RECTAPUR®
: VWR Chemicals