Copper(II) nitrate hemipentahydrate 99.999%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate
CAS Number:19004-19-4
:
Formula:Cu(NO₃)₂·2.5H₂O
MDL Number:MFCD00211298
Molecular Weight:277.63 g/mol
Melting Pt:255 °C
Storage Temperature:Refrigerator
UN:3085
ADR:5.1+8 II
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together