Order Entry
Northern Ireland
ContactUsLinkComponent
Copper(I) bromide dimethyl sulphide complex 99%
Irritant
Copper(I) bromide dimethyl sulphide complex 99%
Catalog # APOSIN1541-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Copper(I) bromide dimethyl sulphide complex 99%
Catalog # APOSIN1541-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Melting Pt:132 °C (dec.)
Storage Temperature:Ambient
UN:3335
ADR:9,III
 :205.58 g/mol
 :GHS07
CAS Number:54678-23-8
Formula:C₂H₆BrCuS
MDL Number:MFCD00043295