Cesium hydroxide 50% in aqueous solution 99,9%
:21351-79-1
:
:CsOH
:MFCD00010964
:149.91 g/mol
:1,72 g/cm³ (25 °C)
:Ambient
:244-344-1
:2681
:8,II
:GHS05
:GHS07
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Cesium hydroxide 50% (w/w) , 99% (metals basis) in aqueous solution
: Thermo Fisher Scientific
Ampoules, standard, WHEATON®
: DWK Life Sciences