

Cerium(IV) sulphate tetrahydrate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10294-42-5
:
Formula:Ce(SO₄)₂·4H₂O
MDL Number:MFCD00149427
Density:3,91 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3085
ADR:5.1+8,III
:404.3 g/mol
:GHS03
:GHS05
Frequently Bought Together