Cerium trichloride hydrate 99,9%
Catalog # APOSIN1414-250G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
Specifications
About this item
Synonyms: Cerium(III) chloride hydrate, Cerous chloride
:
:246.48 g/mol (anhy)
EINECS:232-227-8
:GHS07
Storage Temperature:Ambient
CAS Number:19423-76-8
Formula:CeCl₃·xH₂O
MDL Number:MFCD00149633
Melting Pt:848 °C
Merck Index:12,02041