

Barium nitrate 99,9%
Catalog # APOSIN1204-50G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 g
- Seq:3368
Specifications
About this item
:
Melting Pt:592 °C (dec.)
Density:3,23 g/cm³ (25 °C)
ADR:5.1,II
:261.34 g/mol
UN:1479
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:10022-31-8
Formula:Ba(NO₃)₂
MDL Number:MFCD00003442