Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Tobramycin

Irritant
Supplier:   Apollo Scientific
Tobramycin
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Tobramycin is an aminoglycoside antibiotic and hasa bactericidal action against many gram-negative bacteria. Aminoglycosides are taken into sensitivebacterial cells by an active transport process.

Synonyms:  (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
  • CAS Number:32986-56-4
  • Formula:C₁₈H₃₇N₅O₉
  • MDL Number:MFCD00077885
  • Molecular Weight:467.51 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:251-322-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - £105.00
    £105.00
    1 * 500 mg
       £105.00

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £184.00
    £184.00
    1 * 1 g
       £184.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.