
Octyl-β-D-thioglucopyranoside ≥99%, Ultrapure
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85618-21-9
:
Formula:C₁₄H₂₈O₅S
MDL Number:MFCD00012189
:308.44 g/mol
Melting Pt:125…131 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together