
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99.5% low sodium
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1132-61-2
:
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
:209.27 g/mol
EINECS:214-478-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together