Folic acid ≥98%
Catalog # APOSBIF0608-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:441.41 g/mol
EINECS:200-419-0
:GHS07
Storage Temperature:Ambient
CAS Number:59-30-3
Formula:C₁₉H₁₉N₇O₆
MDL Number:MFCD00079305