18-Lupen-3-one
Catalog # APOSBIDF1010-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:424.71 g/mol
Melting Pt:165…167 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1617-70-5
Formula:C₃₀H₄₈O
MDL Number:MFCD00083578