
Triptolide
Catalog # APOSBIA8418-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:360.41 g/mol
Melting Pt:235…237 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:38748-32-2
Formula:C₂₀H₂₄O₆
MDL Number:MFCD00210565