
Aphidicolin (from Nigrospora sphaerica)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38966-21-1
:
Formula:C₂₀H₃₄O₄
MDL Number:MFCD00083214
:338.488 g/mol
Melting Pt:224…233 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together